• USA Home
  • Search Results

  • 2naphthoicacid172189309411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Naphthoic acid

2 Products
Synonym:   2-Naphthalenecarboxylic acid
Image of 2-Naphthoic acid
CAS Number:
Linear Formula:
 C10H7CO2H
Molecular Weight:
 172.18
 Beilstein/REAXYS Number:
 972039
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More