• USA Home
  • Search Results

  • 2naphthol1441713519311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Naphthol

5 Products
Synonym:   β-Naphthol, 2-Hydroxynaphthalene
Image of 2-Naphthol
CAS Number:
Linear Formula:
 C10H7OH
Molecular Weight:
 144.17
 Beilstein/REAXYS Number:
 742134
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39023410
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Potency
  Fluorescence Test
  Special Grade
  Melting Point
Show More