• USA Home
  • Search Results

  • 2naphthylamine143199159811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Naphthylamine

2 Products
Synonym:   β-Naphthylamine, 2-Aminonaphthalene
Image of 2-Naphthylamine
CAS Number:
Linear Formula:
 C10H7NH2
Molecular Weight:
 143.19
 Beilstein/REAXYS Number:
 3939429
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More