• USA Home
  • Search Results

  • 2naphthylphosphatedisodiumsalt268113168198811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Naphthyl phosphate disodium salt

1 Products
Image of 2-Naphthyl phosphate disodium salt
CAS Number:
Linear Formula:
 C10H7OP(O)(ONa)2
Molecular Weight:
 268.11
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More