• USA Home
  • Search Results

  • 2nitro1propanol10509290296711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Nitro-1-propanol

1 Products
Image of 2-Nitro-1-propanol
CAS Number:
Linear Formula:
 CH3CH(NO2)CH2OH
Molecular Weight:
 105.09
 Beilstein/REAXYS Number:
 1748592
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More