• USA Home
  • Search Results

  • 2nitrophenol139118875511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Nitrophenol

2 Products
Image of 2-Nitrophenol
CAS Number:
Linear Formula:
 O2NC6H4OH
Molecular Weight:
 139.11
 Beilstein/REAXYS Number:
 775403
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More