• USA Home
  • Search Results

  • 2o2acetamido2deoxybdglucopyranosyldmannose383353462173311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-O-(2-Acetamido-2-deoxy-β-D-glucopyranosyl)-D-mannose

1 Products
Synonym:   β-D-GlcNAc-(1→2)-D-Man, N-Acetylglucosaminyl-β-1,2-mannose
Image of 2-<I>O</I>-<WBR>(2-<WBR>Acetamido-<WBR>2-<WBR>deoxy-β-<SC>D</SC>-glucopyranosyl)-<SC>D</SC>-mannose
CAS Number:
Empirical Formula (Hill Notation):
 C14H25NO11
Molecular Weight:
 383.35
 NACRES
 NA.25
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More