• USA Home
  • Search Results

  • 2octanone1282111113711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Octanone

4 Products
Synonym:   Hexyl methyl ketone
Image of 2-Octanone
CAS Number:
Linear Formula:
 CH3(CH2)5COCH3
Molecular Weight:
 128.21
 Beilstein/REAXYS Number:
 635843
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Assay
  Agency/Method
Show More