• USA Home
  • Search Results

  • 2octyl1dodecanol29855533342611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Octyl-1-dodecanol

1 Products
Synonym:   Octyldodecanol
Image of 2-Octyl-1-dodecanol
CAS Number:
Linear Formula:
 CH3(CH2)9CH[(CH2)7CH3]CH2OH
Molecular Weight:
 298.55
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
464481 2-Octyl-1-dodecanol, 97%,