• USA Home
  • Search Results

  • 2octyn1ol126202073958611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Octyn-1-ol

1 Products
Image of 2-Octyn-1-ol
CAS Number:
Linear Formula:
 CH3(CH2)4C≡COH
Molecular Weight:
 126.20
 Beilstein/REAXYS Number:
 1744120
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
630837 2-Octyn-1-ol, 97%,