• USA Home
  • Search Results

  • 2pentylfuran13821377769311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Pentylfuran

2 Products
Synonym:   2-Amylfuran
Image of 2-Pentylfuran
CAS Number:
Empirical Formula (Hill Notation):
 C9H14O
Molecular Weight:
 138.21
 Council of Europe no.:
 10966
 EC Number:
 FEMA Number:
 3317
 Flavis number:
 13.059
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Assay
  Agency/Method
Show More