• USA Home
  • Search Results

  • 2pentyne681262721411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Pentyne

1 Products
Image of 2-Pentyne
CAS Number:
Linear Formula:
 CH3C≡CCH2CH3
Molecular Weight:
 68.12
 Beilstein/REAXYS Number:
 1696938
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
271357 2-Pentyne, 98%,