• USA Home
  • Search Results

  • 2phenoxyethanol1381612299611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Phenoxyethanol

3 Products
Synonym:   Ethylene glycol monophenyl ether, PHE-G, PHE-S, PHE, PhG, Phenylglycol
Image of 2-Phenoxyethanol
CAS Number:
Linear Formula:
 C6H5OCH2CH2OH
Molecular Weight:
 138.16
 Beilstein/REAXYS Number:
 1364011
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Refractive Index
  Anal. Standard Form
  Assay
  Special Grade
  Physical Form
Show More