• USA Home
  • Search Results

  • 2phenyl1propanol13619112385911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Phenyl-1-propanol

2 Products
Synonym:   β-Methylphenethyl alcohol, (±)-2-Phenyl-1-propanol
Image of 2-Phenyl-1-propanol
CAS Number:
Linear Formula:
 CH3CH(C6H5)CH2OH
Molecular Weight:
 136.19
 Beilstein/REAXYS Number:
 1906760
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
  Special Grade
Show More