• USA Home
  • Search Results

  • 2phenylethanol122166012811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Phenylethanol

2 Products
Synonym:   β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA
Image of 2-Phenylethanol
CAS Number:
Linear Formula:
 C6H5CH2CH2OH
Molecular Weight:
 122.16
 Beilstein/REAXYS Number:
 1905732
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More