• USA Home
  • Search Results

  • 2phenylphenol170219043711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Phenylphenol

3 Products
Synonym:   2-Hydroxybiphenyl
Image of 2-Phenylphenol
CAS Number:
Linear Formula:
 C6H5C6H4OH
Molecular Weight:
 170.21
 Beilstein/REAXYS Number:
 606907
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Assay
Show More