• USA Home
  • Search Results

  • 2picoline931310906811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Picoline

1 Products
Synonym:   α-Picoline, 2-Methylpyridine, NSC 3409
Image of 2-Picoline
CAS Number:
Empirical Formula (Hill Notation):
 C6H7N
Molecular Weight:
 93.13
 Beilstein/REAXYS Number:
 104581
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More