• USA Home
  • Search Results

  • 2propanethiol76167533211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Propanethiol

6 Products
Synonym:   Isopropyl mercaptan
Image of 2-Propanethiol
CAS Number:
Linear Formula:
 (CH3)2CHSH
Molecular Weight:
 76.16
 EC Index Number
 200-861-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Biological Source
  Special Grade
Show More