• USA Home
  • Search Results

  • 2propanol60106763011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Propanol

31 Products
Synonym:   sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
Image of 2-Propanol
CAS Number:
Linear Formula:
 (CH3)2CHOH
Molecular Weight:
 60.10
 Beilstein/REAXYS Number:
 635639
 EC Number:
 MDL number:
 NACRES:
 NA.52
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Quality clarifier
  Density
  Anal. Standard Form
  UV-Absorption Condition
  Restriction
  Suitability
  Purity Grade
  Transmittance
  Product Line
  Vapor pressure
  Refractive Index
  Transition temperature
  Agency/Method
  Color
  Potency
  Solubility
  pH Value
  Application(s)
  Special Grade
  UV-Absorption Values
  Physical Form
Show More

Product #
Description
I9516 2-Propanol, BioReagent, for molecular biology, ≥99.5%,
34863 2-Propanol, suitable for HPLC, 99.9%,
190764 2-Propanol, ACS reagent, ≥99.5%,
278475 2-Propanol, anhydrous, 99.5%,
33539 2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC),
650447 2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%,
1.02781 2-Propanol, hypergrade for LC-MS LiChrosolv®,
59304 2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC),
109634 2-Propanol, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
109827 2-Propanol, Laboratory Reagent, ≥99.5%,
733458 2-Propanol, electronic grade, 99.999% trace metals basis,
59300 2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC),
PHR1072 2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material,
91237 2-Propanol, analytical standard,
1.01040 2-Propanol, gradient grade for liquid chromatography LiChrosolv®,
24137 2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC),
1.00995 2-Propanol, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,
1570428 2-Propanol, United States Pharmacopeia (USP) Reference Standard,
437522 2-Propanol, ACS reagent, ≥99.5%,
439207 2-Propanol, suitable for HPLC, 99.5%,
1.00993 2-Propanol, for spectroscopy Uvasol®,
1.07022 2-Propanol, for analysis EMPARTA® ACS,
8.18766 2-Propanol, EMPLURA®,
1.00998 2-Propanol, for gas chromatography ECD and FID SupraSolv®,
675431 2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles,
1.13350 2-Propanol, for preparative chromatography Prepsolv,
1.00272 2-Propanol, standard for the analysis of tobacco,
909955 2-Propanol, BioRenewable, ReagentPlus®, ≥99.5%, Green Alternative
1.05008 2-Propanol, suitable for preparative chromatography EMPROVE® EXPERT Ph Eur,BP,USP,
673773 2-Propanol, ACS reagent, ≥99.5%,
270490 2-Propanol, 99.5%, HPLC grade,