• USA Home
  • Search Results

  • 2r3r3bocamino2hydroxy4phenylbutyricacid295337717141611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

1 Products
Synonym:   Boc-(2R,3R)-3-amino-2-hydroxy-4-phenylbutyric acid
Image of (2<I>R</I>,3<I>R</I>)<WBR>-<WBR>3-<WBR>(Boc-<WBR>amino)<WBR>-<WBR>2-<WBR>hydroxy-<WBR>4-<WBR>phenylbutyric acid
CAS Number:
Empirical Formula (Hill Notation):
 C15H21NO5
Molecular Weight:
 295.33
 Beilstein/REAXYS Number:
 8993437
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More