• USA Home
  • Search Results

  • 2s3r4sacarboxycyclopropylglycine1591411785795111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(2S,3R,4S)-α-(Carboxycyclopropyl)glycine

1 Products
Synonym:   (2S,1′R,2′S)-2-(2-Carboxycyclopropyl)glycine, (2S,3R,4S)-CCG, L-CCG-IV
Image of (2S,3R,4S)-α-(Carboxycyclopropyl)glycine
CAS Number:
Empirical Formula (Hill Notation):
 C6H9NO4
Molecular Weight:
 159.14
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More