• USA Home
  • Search Results

  • 2secbutyl3methoxypyrazine166222416870511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-sec-Butyl-3-methoxypyrazine

1 Products
Image of 2-<I>sec</I>-<WBR>Butyl-<WBR>3-<WBR>methoxypyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C9H14N2O
Molecular Weight:
 166.22
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More