• USA Home
  • Search Results

  • 2tertbutyl65chloro2hbenzotriazol2yl4methylphenol31580389611511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol

2 Products
Synonym:   2-(2-Hydroxy-3-tert -butyl-5-methylphenyl)-5-chloro-2H -benzotriazole, 2-(3-tert -Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H -benzotriazole, 2-(5-Chloro-2H -benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol, 2-(5-Chloro-2-benzotriazolyl)-6-tert -butyl-p -cresol, 2-[5-Chloro-2H -benzotriazol-2-yl]-4-methyl-6-(tert -butyl)phenol
Image of 2-<I>tert</I>-Butyl-6-(5-chloro-2<I>H</I>-<WBR>benzotriazol-<WBR>2-<WBR>yl)<WBR>-<WBR>4-<WBR>methylphenol
CAS Number:
Empirical Formula (Hill Notation):
 C17H18ClN3O
Molecular Weight:
 315.80
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More