• USA Home
  • Search Results

  • 2tertbutylaminoethanol11719462070611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-tert-Butylamino-ethanol

1 Products
Synonym:   N-tert-Butylethanolamin, N-tert-Butylethanolamin
Image of 2-<WBR>tert-<WBR>Butylamino-<WBR>ethanol
CAS Number:
Linear Formula:
 (CH3)3CNHCH2CH2OH
Molecular Weight:
 117.19
 EC Index Number
 225-034-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More