• USA Home
  • Search Results

  • 2trimethylsilyl13dithiane192421341142211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-(Trimethylsilyl)-1,3-dithiane

1 Products
Image of 2-<WBR>(Trimethylsilyl)<WBR>-<WBR>1,3-<WBR>dithiane
CAS Number:
Empirical Formula (Hill Notation):
 C7H16S2Si
Molecular Weight:
 192.42
 Beilstein/REAXYS Number:
 1616463
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More