• USA Home
  • Search Results

  • 335triiodolthyroninesodiumsalt672965506111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3′,5-Triiodo-L-thyronine sodium salt

2 Products
Synonym:   Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3
Image of 3,3′,5-Triiodo-<SC>L</SC>-thyronine sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C15H11I3NNaO4
Molecular Weight:
 672.96
 Beilstein/REAXYS Number:
 8179867
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More