• USA Home
  • Search Results

  • 33cholamidopropyldimethylammonio1propanesulfonatehydrate61488anhydrousbasis33171745411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate

1 Products
Synonym:   CHAPS
Image of 3-<WBR>[(3-<WBR>Cholamidopropyl)<WBR>dimethylammonio]<WBR>-<WBR>1-<WBR>propanesulfonate hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C32H58N2O7S · xH2O
Molecular Weight:
 614.88 (anhydrous basis)
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More