• USA Home
  • Search Results

  • 33diethoxy1propyne128171016087911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3-Diethoxy-1-propyne

1 Products
Synonym:   Propargylaldehyde diethyl acetal, Propiolaldehyde diethyl acetal
Image of 3,3-<WBR>Diethoxy-<WBR>1-<WBR>propyne
CAS Number:
Linear Formula:
 (C2H5O)2CHC≡CH
Molecular Weight:
 128.17
 Beilstein/REAXYS Number:
 1701566
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More