• USA Home
  • Search Results

  • 33diindolylmethane24631196805411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3′-Diindolylmethane

2 Products
Synonym:   3,3′-Bisindolylmethane, DIM
Image of 3,3′-Diindolylmethane
CAS Number:
Empirical Formula (Hill Notation):
 C17H14N2
Molecular Weight:
 246.31
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More