• USA Home
  • Search Results

  • 33dimethyl1butyne821491792011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3-Dimethyl-1-butyne

1 Products
Synonym:   tert-Butylacetylene
Image of 3,3-<WBR>Dimethyl-<WBR>1-<WBR>butyne
CAS Number:
Linear Formula:
 (CH3)3CC≡CH
Molecular Weight:
 82.14
 Beilstein/REAXYS Number:
 1697100
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More