• USA Home
  • Search Results

  • 33methylenebis4hydroxycoumarin336296676211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3′-Methylene-bis(4-hydroxycoumarin)

1 Products
Synonym:   Bishydroxycoumarin, Dicoumarol, Dicumarol
Image of 3,3′-Methylene-bis(4-hydroxycoumarin)
CAS Number:
Empirical Formula (Hill Notation):
 C19H12O6
Molecular Weight:
 336.29
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More