• USA Home
  • Search Results

  • 33thiodipropionicacid1782111117111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,3′-Thiodipropionic acid

3 Products
Synonym:   Bis(2-carboxyethyl) sulfide
Image of 3,3′-Thiodipropionic acid
CAS Number:
Linear Formula:
 S(CH2CH2COOH)2
Molecular Weight:
 178.21
 Beilstein/REAXYS Number:
 1210299
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More