• USA Home
  • Search Results

  • 3456tetrachloro12benzoquinone24588243553211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4,5,6-Tetrachloro-1,2-benzoquinone

1 Products
Synonym:   o-Chloranil
Image of 3,4,5,6-<WBR>Tetrachloro-<WBR>1,2-<WBR>benzoquinone
CAS Number:
Linear Formula:
 C6Cl4(=O)2
Molecular Weight:
 245.88
 Beilstein/REAXYS Number:
 1309782
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More