• USA Home
  • Search Results

  • 345trichloroaniline1964663491311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4,5-Trichloroaniline

1 Products
Image of 3,4,5-<WBR>Trichloroaniline
CAS Number:
Linear Formula:
 Cl3C6H2NH2
Molecular Weight:
 196.46
 Beilstein/REAXYS Number:
 638880
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More