• USA Home
  • Search Results

  • 34dehydrolproline11311404388311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Dehydro-L-proline

1 Products
Synonym:   (S)-3-Pyrroline-2-carboxylic acid, 3,4-Didehydro-L-proline
Image of 3,4-Dehydro-<SC>L</SC>-proline
CAS Number:
Empirical Formula (Hill Notation):
 C5H7NO2
Molecular Weight:
 113.11
 Beilstein/REAXYS Number:
 5376764
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More

Product #
Description
D4893 3,4-Dehydro-L-proline,