• USA Home
  • Search Results

  • 34diaminopyridine109135496611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Diaminopyridine

2 Products
Image of 3,4-Diaminopyridine
CAS Number:
Empirical Formula (Hill Notation):
 C5H7N3
Molecular Weight:
 109.13
 Beilstein/REAXYS Number:
 110232
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
Show More