• USA Home
  • Search Results

  • 34dibromohexane243971623028711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Dibromohexane

1 Products
Image of 3,4-Dibromohexane
CAS Number:
Linear Formula:
 C2H5CH(Br)CH(Br)C2H5
Molecular Weight:
 243.97
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
446610 3,4-Dibromohexane, 98%,