• USA Home
  • Search Results

  • 34difluorophenylaceticacid1721365893511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Difluorophenylacetic acid

1 Products
Image of 3,4-<WBR>Difluorophenylacetic acid
CAS Number:
Linear Formula:
 F2C6H3CH2CO2H
Molecular Weight:
 172.13
 Beilstein/REAXYS Number:
 2577768
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More