• USA Home
  • Search Results

  • 34dihydroxybenzaldehyde1381213985511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Dihydroxybenzaldehyde

4 Products
Synonym:   Protocatechualdehyde
Image of 3,4-<WBR>Dihydroxybenzaldehyde
CAS Number:
Linear Formula:
 (HO)2C6H3CHO
Molecular Weight:
 138.12
 Beilstein/REAXYS Number:
 774381
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
  Melting Point
Show More