• USA Home
  • Search Results

  • 34dihydroxylphenylalanine197195992711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Dihydroxy-L-phenylalanine

2 Products
Synonym:   3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa
Image of 3,4-Dihydroxy-<SC>L-</SC>phenylalanine
CAS Number:
Linear Formula:
 (HO)2C6H3CH2CH(NH2)CO2H
Molecular Weight:
 197.19
 Beilstein/REAXYS Number:
 2215169
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More