• USA Home
  • Search Results

  • 34dimethylphenol122169565811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Dimethylphenol

2 Products
Synonym:   3,4-DMP, asym.-o-Xylenol, 3,4-Dimethylphenol
Image of 3,4-Dimethylphenol
CAS Number:
Linear Formula:
 3,4-(CH3)2C6H3OH
Molecular Weight:
 122.16
 EC Index Number
 202-439-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Melting Point
  Titertest
Show More