• USA Home
  • Search Results

  • 34xylenol122169565811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,4-Xylenol

1 Products
Synonym:   3,4-Dimethylphenol, 4-Hydroxy-o-xylene
Image of 3,4-Xylenol
CAS Number:
Linear Formula:
 (CH3)2C6H3OH
Molecular Weight:
 122.16
 Beilstein/REAXYS Number:
 1099267
 Council of Europe no.:
 11262
 EC Number:
 FEMA Number:
 3596
 Flavis number:
 4.048
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More

Product #
Description
W359602 3,4-Xylenol, ≥98%, FG,