• USA Home
  • Search Results

  • 35diamino124triazole9909145577211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Diamino-1,2,4-triazole

1 Products
Synonym:   Guanazole
Image of 3,5-<WBR>Diamino-<WBR>1,2,4-<WBR>triazole
CAS Number:
Empirical Formula (Hill Notation):
 C2H5N5
Molecular Weight:
 99.09
 Beilstein/REAXYS Number:
 112467
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More