• USA Home
  • Search Results

  • 35dicaffeoylquinicacid51645245053511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Di-caffeoylquinic acid

2 Products
Image of 3,5-<WBR>Di-<WBR>caffeoylquinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C25H24O12
Molecular Weight:
 516.45
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More