• USA Home
  • Search Results

  • 35diiodoltyrosinedihydrate469011883559111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Diiodo-L-tyrosine dihydrate

1 Products
Image of 3,5-Diiodo-<SC>L</SC>-tyrosine dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C9H9I2NO3 · 2H2O
Molecular Weight:
 469.01
 Beilstein/REAXYS Number:
 2218691
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More