• USA Home
  • Search Results

  • 35dimethoxybenzaldehyde16617731134411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Dimethoxybenzaldehyde

1 Products
Image of 3,5-<WBR>Dimethoxybenzaldehyde
CAS Number:
Linear Formula:
 (CH3O)2C6H3CHO
Molecular Weight:
 166.17
 Beilstein/REAXYS Number:
 2042374
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More