• USA Home
  • Search Results

  • 35dinitrobenzylalcohol198137102243011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Dinitrobenzyl alcohol

1 Products
Image of 3,5-Dinitrobenzyl alcohol
CAS Number:
Linear Formula:
 (O2N)2C6H3CH2OH
Molecular Weight:
 198.13
 Beilstein/REAXYS Number:
 2054388
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More