• USA Home
  • Search Results

  • 35heptanedione12817742454611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3,5-Heptanedione

1 Products
Image of 3,5-Heptanedione
CAS Number:
Linear Formula:
 H3CCH2COCH2COCH2CH3
Molecular Weight:
 128.17
 Beilstein/REAXYS Number:
 635979
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
280070 3,5-Heptanedione, 97%,