• USA Home
  • Search Results

  • 3acetyldeoxynivalenol338355072238811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Acetyldeoxynivalenol

2 Products
Synonym:   3α-Acetoxy-7α,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 3α-Acetylvomitoxin, 3-AcDON
Image of 3-<WBR>Acetyldeoxynivalenol
CAS Number:
Empirical Formula (Hill Notation):
 C17H22O7
Molecular Weight:
 338.35
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More