• USA Home
  • Search Results

  • 3amino12propanediol911161630811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-3-Amino-1,2-propanediol

2 Products
Image of (±)<WBR>-<WBR>3-<WBR>Amino-<WBR>1,2-<WBR>propanediol
CAS Number:
Linear Formula:
 NH2CH2CH(OH)CH2OH
Molecular Weight:
 91.11
 Beilstein/REAXYS Number:
 1719121
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More